Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc20a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016904.2 | NP_001016904.1 | 9 - 10 | 2083 | 64 | |
XM_012962996.2 | XP_012818450.1 | 9 - 10 | 2111 | 64 | ||
XM_012963000.2 | XP_012818454.1 | 9 - 10 | 2087 | 64 | ||
GenBank mRNA | CR848513.2 | - | 9 - 10 | 2083 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Xenopus tropicalis Probe spans exons |