Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccnyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016919.2 | NP_001016919.1 | 3 - 4 | 324 | 103 | |
GenBank mRNA | BC161467.1 | - | 3 - 4 | 509 | 103 | |
CR848188.2 | - | 3 - 4 | 324 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |