Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016961.3 | NP_001016961.2 | 1 - 2 | 363 | 95 | |
GenBank mRNA | BC127345.1 | - | 1 - 2 | 363 | 95 | |
CR761361.2 | - | 1 - 2 | 465 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |