Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nedd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016973.2 | NP_001016973.1 | 2 - 3 | 91 | 96 | |
GenBank mRNA | BC122002.1 | - | 2 - 3 | 76 | 96 | |
CR848418.2 | - | 2 - 3 | 91 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |