Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flvcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016982.2 | NP_001016982.1 | 7 - 8 | 1301 | 97 | |
XM_012968451.2 | XP_012823905.1 | 7 - 8 | 1889 | 97 | ||
XM_012968452.2 | XP_012823906.1 | 7 - 8 | 1549 | 97 | ||
XM_012968453.2 | XP_012823907.1 | 7 - 8 | 1489 | 97 | ||
XM_012968454.2 | XP_012823908.1 | 7 - 8 | 1481 | 97 | ||
XM_012968455.2 | XP_012823909.1 | 7 - 8 | 1892 | 97 | ||
XM_012968456.2 | XP_012823910.1 | 7 - 8 | 1890 | 97 | ||
XM_012968457.2 | XP_012823911.1 | 7 - 8 | 1892 | 97 | ||
XM_018096266.1 | XP_017951755.1 | 7 - 8 | 1894 | 97 | ||
XM_018096267.1 | XP_017951756.1 | 7 - 8 | 1896 | 97 | ||
XM_018096268.1 | XP_017951757.1 | 7 - 8 | 1898 | 97 | ||
XM_018096269.1 | XP_017951758.1 | 7 - 8 | 1898 | 97 | ||
GenBank mRNA | BC122001.1 | - | 7 - 8 | 1286 | 97 | |
CR848328.2 | - | 7 - 8 | 1301 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |