Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017043.2 | NP_001017043.1 | 4 - 5 | 575 | 96 | |
XM_012968462.2 | XP_012823916.1 | 4 - 5 | 627 | 96 | ||
GenBank mRNA | CR761859.2 | - | 4 - 5 | 575 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Xenopus tropicalis Probe spans exons |