Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrps30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017073.2 | NP_001017073.1 | 4 - 5 | 1040 | 74 | |
GenBank mRNA | CR761538.2 | - | 4 - 5 | 1040 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Xenopus tropicalis Probe spans exons |