Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp1r8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017079.2 | NP_001017079.1 | 3 - 4 | 273 | 84 | |
XM_012957029.2 | XP_012812483.1 | 3 - 4 | 342 | 84 | ||
GenBank mRNA | BC135351.1 | - | 3 - 4 | 275 | 84 | |
CR761503.2 | - | 3 - 4 | 273 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |