Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp1r8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017079.2 | NP_001017079.1 | 4 - 5 | 484 | 95 | |
XM_012957029.2 | XP_012812483.1 | 4 - 5 | 553 | 95 | ||
GenBank mRNA | BC135351.1 | - | 4 - 5 | 486 | 95 | |
CR761503.2 | - | 4 - 5 | 484 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |