Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017080.2 | NP_001017080.1 | 3 - 4 | 687 | 75 | |
XM_018092105.1 | XP_017947594.1 | 4 - 5 | 870 | 75 | ||
GenBank mRNA | BC131830.1 | - | 4 - 5 | 713 | 75 | |
BC167997.1 | - | 3 - 4 | 631 | 75 | ||
CR761485.2 | - | 3 - 4 | 687 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |