Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017103.3 | NP_001017103.2 | 1 - 2 | 619 | 95 | |
XM_012959080.1 | XP_012814534.1 | 1 - 2 | 686 | 95 | ||
GenBank mRNA | BC136173.1 | - | 1 - 2 | 619 | 95 | |
CR761277.2 | - | 1 - 2 | 648 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |