Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gng7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017133.2 | NP_001017133.1 | 1 - 2 | 135 | 81 | |
GenBank mRNA | BC168546.1 | - | 2 - 3 | 175 | 81 | |
CR761071.2 | - | 1 - 2 | 135 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |