Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017135.2 | NP_001017135.1 | 2 - 3 | 197 | 67 | |
XM_012970449.1 | XP_012825903.1 | 2 - 3 | 245 | 67 | ||
XM_012970450.2 | XP_012825904.1 | 2 - 3 | 243 | 67 | ||
GenBank mRNA | BC153688.1 | - | 1 - 2 | 167 | 67 | |
BC167337.1 | - | 2 - 3 | 186 | 67 | ||
CR761043.2 | - | 2 - 3 | 197 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |