Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017138.2 | NP_001017138.1 | 2 - 3 | 292 | 86 | |
GenBank mRNA | BC157272.1 | - | 2 - 3 | 278 | 86 | |
CR761012.2 | - | 2 - 3 | 292 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |