Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfand3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017147.2 | NP_001017147.1 | 375 | 85 | ||
GenBank mRNA | BC157766.1 | - | 419 | 85 | ||
CR760946.2 | - | 375 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |