Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017150.2 | NP_001017150.1 | 3 - 4 | 465 | 67 | |
GenBank mRNA | CR760924.2 | - | 3 - 4 | 465 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Xenopus tropicalis Probe spans exons |