Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017173.2 | NP_001017173.1 | 5 - 6 | 606 | 87 | |
XM_012970452.2 | XP_012825906.1 | 4 - 5 | 799 | 87 | ||
GenBank mRNA | BC121599.1 | - | 4 - 5 | 406 | 87 | |
BC171137.1 | - | 4 - 5 | 653 | 87 | ||
CR760741.2 | - | 5 - 6 | 606 | 87 |
Gene Symbol: rps6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC161304.1 | - | 1341 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |