Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017198.2 | NP_001017198.1 | 3 - 4 | 155 | 102 | |
XM_018097130.1 | XP_017952619.1 | 3 - 4 | 274 | 102 | ||
GenBank mRNA | CR760538.2 | - | 3 - 4 | 155 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Xenopus tropicalis Probe spans exons |