Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qpct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017245.2 | NP_001017245.1 | 6 - 7 | 952 | 91 | |
GenBank mRNA | BC118705.1 | - | 6 - 7 | 996 | 91 | |
CR760147.2 | - | 6 - 7 | 952 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |