Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017262.2 | NP_001017262.1 | 2 - 3 | 352 | 76 | |
GenBank mRNA | BC155974.1 | - | 2 - 3 | 407 | 76 | |
BC158237.1 | - | 2 - 3 | 407 | 76 | ||
CR760005.2 | - | 2 - 3 | 352 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |