Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fto | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017273.2 | NP_001017273.1 | 7 - 8 | 1271 | 183 | |
XM_012961194.2 | XP_012816648.1 | 7 - 8 | 1288 | 183 | ||
XM_018093066.1 | XP_017948555.1 | 7 - 8 | 1288 | 183 | ||
XM_018093067.1 | XP_017948556.1 | 6 - 7 | 1156 | 183 | ||
XM_018093068.1 | XP_017948557.1 | 8 - 9 | 1058 | 183 | ||
XM_018093069.1 | XP_017948558.1 | 6 - 7 | 1156 | 183 | ||
GenBank mRNA | CR855775.2 | - | 7 - 8 | 1271 | 183 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Xenopus tropicalis Probe spans exons |