Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abhd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017287.2 | NP_001017287.1 | 5 - 6 | 779 | 70 | |
GenBank mRNA | CR855695.2 | - | 5 - 6 | 779 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Xenopus tropicalis Probe spans exons |