Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klhl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017289.2 | NP_001017289.1 | 1 - 2 | 296 | 112 | |
XM_012953592.2 | XP_012809046.1 | 2 - 3 | 455 | 112 | ||
XM_018093771.1 | XP_017949260.1 | 2 - 3 | 1772 | 112 | ||
GenBank mRNA | BC167576.1 | - | 2 - 3 | 339 | 112 | |
CR855674.2 | - | 1 - 2 | 296 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |