Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klhl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017289.2 | NP_001017289.1 | 7 - 8 | 1615 | 82 | |
XM_012953592.2 | XP_012809046.1 | 7 - 8 | 1630 | 82 | ||
XM_018093771.1 | XP_017949260.1 | 8 - 9 | 3091 | 82 | ||
XM_018093773.1 | XP_017949262.1 | 7 - 8 | 1404 | 82 | ||
GenBank mRNA | BC167576.1 | - | 8 - 9 | 1658 | 82 | |
CR855674.2 | - | 7 - 8 | 1615 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |