Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rffl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017296.3 | NP_001017296.2 | 2 - 3 | 263 | 88 | |
XM_012957057.2 | XP_012812511.1 | 2 - 3 | 284 | 88 | ||
XM_018091282.1 | XP_017946771.1 | 3 - 4 | 388 | 88 | ||
GenBank mRNA | BC090590.1 | - | 2 - 3 | 263 | 88 | |
CR855622.2 | - | 2 - 3 | 292 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |