Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018091310.1 | XP_017946799.1 | 2 - 3 | 1026 | 67 | |
GenBank mRNA | BC135209.1 | - | 1 - 2 | 555 | 67 | |
DQ124206.1 | - | 1 - 2 | 726 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |