Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037262.1 | NP_001032339.1 | 3 | 414 | 69 | |
GenBank mRNA | BC087765.1 | - | 3 | 384 | 69 | |
BC157253.1 | - | 3 | 408 | 69 | ||
CR761131.2 | - | 3 | 414 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Both primers and probe map within a single exon |