Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037272.1 | NP_001032349.1 | 5 - 6 | 521 | 85 | |
GenBank mRNA | BC077668.1 | - | 5 - 6 | 526 | 85 | |
BC088002.1 | - | 5 - 6 | 521 | 85 | ||
CR848186.2 | - | 5 - 6 | 470 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |