Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nolc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039730.1 | NP_001034819.1 | 4 - 5 | 443 | 95 | |
XM_012966311.1 | XP_012821765.1 | 4 - 5 | 772 | 95 | ||
XM_012966312.1 | XP_012821766.1 | 4 - 5 | 772 | 95 | ||
XM_018095326.1 | XP_017950815.1 | 4 - 5 | 439 | 95 | ||
GenBank mRNA | BC064870.1 | - | 4 - 5 | 417 | 95 | |
BC077061.1 | - | 4 - 5 | 449 | 95 | ||
BC121842.1 | - | 4 - 5 | 484 | 95 | ||
CR848207.2 | - | 4 - 5 | 443 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |