Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nolc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039730.1 | NP_001034819.1 | 9 - 10 | 1049 | 66 | |
XM_012966311.1 | XP_012821765.1 | 9 - 10 | 1378 | 66 | ||
XM_012966312.1 | XP_012821766.1 | 9 - 10 | 1378 | 66 | ||
XM_018095326.1 | XP_017950815.1 | 9 - 10 | 1045 | 66 | ||
GenBank mRNA | BC064870.1 | - | 9 - 10 | 1023 | 66 | |
BC121842.1 | - | 9 - 10 | 1090 | 66 | ||
CR848207.2 | - | 9 - 10 | 1049 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |