Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnppd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203604.2 | NP_988935.2 | 7 - 8 | 834 | 80 | |
XM_012970235.2 | XP_012825689.1 | 8 - 9 | 1052 | 80 | ||
GenBank mRNA | BC061409.1 | - | 7 - 8 | 780 | 80 | |
CR848649.2 | - | 7 - 8 | 834 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |