Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203937.3 | NP_989268.3 | 2 - 3 | 382 | 75 | |
GenBank mRNA | BC064215.1 | - | 2 - 3 | 374 | 75 | |
CR760996.2 | - | 2 - 3 | 365 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |