Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp1cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213670.2 | NP_998835.1 | 1 - 2 | 224 | 99 | |
GenBank mRNA | BC067911.1 | - | 1 - 2 | 111 | 99 | |
CR761289.1 | - | 1 - 2 | 224 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |