Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006887.4 | NP_001006888.1 | 2 - 3 | 182 | 71 | |
GenBank mRNA | BC077687.1 | - | 2 - 3 | 183 | 71 | |
CR926191.2 | - | 125 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |