Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: casp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015715.2 | NP_001015715.2 | 10 - 11 | 1637 | 104 | |
GenBank mRNA | BC089646.1 | - | 1525 | 104 | ||
CR942647.2 | - | 10 - 11 | 1637 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |