Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: casp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015715.2 | NP_001015715.2 | 5 - 6 | 870 | 103 | |
GenBank mRNA | BC089646.1 | - | 758 | 103 | ||
CR942647.2 | - | 5 - 6 | 870 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |