Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coq7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016068.3 | NP_001016068.2 | 3 - 4 | 408 | 66 | |
GenBank mRNA | BC091075.1 | - | 3 - 4 | 381 | 66 | |
BC118749.1 | - | 3 - 4 | 408 | 66 | ||
CR762257.2 | - | 3 - 4 | 471 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |