Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b4galt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016211.3 | NP_001016211.1 | 1 - 2 | 300 | 78 | |
XM_012964682.1 | XP_012820136.1 | 2 - 3 | 267 | 78 | ||
GenBank mRNA | BC122889.1 | - | 1 - 2 | 300 | 78 | |
CR926330.2 | - | 2 - 3 | 240 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |