Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: diablo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016281.3 | NP_001016281.1 | 1 - 2 | 47 | 78 | |
XM_018093111.1 | XP_017948600.1 | 2 - 3 | 350 | 78 | ||
GenBank mRNA | BC121212.1 | - | 1 - 2 | 47 | 78 | |
CR761608.2 | - | 1 - 2 | 66 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |