Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: diablo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016281.3 | NP_001016281.1 | 4 - 5 | 401 | 87 | |
XM_018093111.1 | XP_017948600.1 | 5 - 6 | 704 | 87 | ||
GenBank mRNA | BC121212.1 | - | 4 - 5 | 401 | 87 | |
CR761608.2 | - | 4 - 5 | 420 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |