Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trappc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016642.3 | NP_001016642.1 | 2 - 3 | 138 | 90 | |
XM_012956993.2 | XP_012812447.1 | 2 - 3 | 303 | 90 | ||
GenBank mRNA | BC121995.1 | - | 2 - 3 | 138 | 90 | |
CR760400.2 | - | 1 - 2 | 128 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |