Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elavl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030327.4 | NP_001025498.3 | 4 - 5 | 589 | 104 | |
XM_012963568.2 | XP_012819022.1 | 5 - 6 | 572 | 104 | ||
XM_012963570.1 | XP_012819024.1 | 5 - 6 | 1635 | 104 | ||
XM_012963574.2 | XP_012819028.1 | 4 - 5 | 597 | 104 | ||
XM_012963578.1 | XP_012819032.1 | 4 - 5 | 1632 | 104 | ||
XM_012963582.2 | XP_012819036.1 | 5 - 6 | 572 | 104 | ||
XM_012963585.2 | XP_012819039.1 | 4 - 5 | 584 | 104 | ||
XM_012963590.2 | XP_012819044.1 | 4 - 5 | 481 | 104 | ||
XM_012963597.2 | XP_012819051.1 | 3 - 4 | 493 | 104 | ||
XM_012963602.2 | XP_012819056.1 | 5 - 6 | 572 | 104 | ||
XM_012963604.2 | XP_012819058.1 | 4 - 5 | 633 | 104 | ||
XM_018093799.1 | XP_017949288.1 | 5 - 6 | 563 | 104 | ||
XM_018093803.1 | XP_017949292.1 | 5 - 6 | 569 | 104 | ||
XM_018093807.1 | XP_017949296.1 | 5 - 6 | 638 | 104 | ||
GenBank mRNA | AL859085.2 | - | 4 - 5 | 589 | 104 | |
BC127338.1 | - | 4 - 5 | 558 | 104 | ||
CR761432.2 | - | 3 - 4 | 439 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |