Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lysmd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044403.2 | NP_001037868.1 | 1 - 2 | 475 | 103 | |
GenBank mRNA | BC107589.1 | - | 1 - 2 | 295 | 103 | |
BC118885.1 | - | 1 - 2 | 268 | 103 | ||
CN093402.1 | - | 1 - 2 | 478 | 103 | ||
CU075816.1 | - | 1 - 2 | 264 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |