Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mgarp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002932039.4 | XP_002932085.1 | 2 - 3 | 348 | 78 | |
GenBank mRNA | CR760051.2 | - | 2 - 3 | 186 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Xenopus tropicalis Probe spans exons |