Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc38a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044435.1 | NP_001037900.1 | 7 - 8 | 1224 | 92 | |
XM_012961231.2 | XP_012816685.1 | 8 - 9 | 1025 | 92 | ||
XM_012961232.2 | XP_012816686.1 | 8 - 9 | 1074 | 92 | ||
XM_018093079.1 | XP_017948568.1 | 8 - 9 | 1037 | 92 | ||
GenBank mRNA | CR760593.2 | - | 7 - 8 | 1224 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |