Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044440.1 | NP_001037905.1 | 2 - 3 | 389 | 111 | |
XM_012964032.2 | XP_012819486.1 | 3 - 4 | 491 | 111 | ||
XM_012964035.2 | XP_012819489.1 | 3 - 4 | 478 | 111 | ||
XM_012964039.2 | XP_012819493.1 | 3 - 4 | 438 | 111 | ||
XM_012964044.2 | XP_012819498.1 | 3 - 4 | 423 | 111 | ||
GenBank mRNA | BC166937.1 | - | 3 - 4 | 479 | 111 | |
CR760680.2 | - | 2 - 3 | 389 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |