Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snx31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044449.1 | NP_001037914.1 | 11 - 12 | 1112 | 88 | |
XM_012964825.2 | XP_012820279.1 | 11 - 12 | 1345 | 88 | ||
XM_012964826.2 | XP_012820280.1 | 11 - 12 | 1343 | 88 | ||
XM_012964827.2 | XP_012820281.1 | 11 - 12 | 1349 | 88 | ||
XM_012964828.2 | XP_012820282.1 | 11 - 12 | 1347 | 88 | ||
XM_012964829.2 | XP_012820283.1 | 11 - 12 | 1328 | 88 | ||
XM_012964830.2 | XP_012820284.1 | 11 - 12 | 1326 | 88 | ||
XM_012964831.2 | XP_012820285.1 | 11 - 12 | 1330 | 88 | ||
XM_012964832.2 | XP_012820286.1 | 12 - 13 | 1287 | 88 | ||
XM_012964833.2 | XP_012820287.1 | 11 - 12 | 1151 | 88 | ||
XM_012964835.2 | XP_012820289.1 | 11 - 12 | 1133 | 88 | ||
GenBank mRNA | CR760929.2 | - | 11 - 12 | 1112 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |