Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bola3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044475.1 | NP_001037940.1 | 2 - 3 | 181 | 112 | |
GenBank mRNA | BC135597.1 | - | 2 - 3 | 223 | 112 | |
CR761390.2 | - | 2 - 3 | 181 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |