Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044478.2 | NP_001037943.2 | 4 - 5 | 373 | 60 | |
GenBank mRNA | BC168456.1 | - | 4 - 5 | 373 | 60 | |
CR761411.2 | - | 4 - 5 | 367 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |