Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zc3h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044484.3 | NP_001037949.3 | 2 - 3 | 1370 | 96 | |
XM_018094683.1 | XP_017950172.1 | 2 - 3 | 1491 | 96 | ||
GenBank mRNA | AY887083.1 | - | 2 - 3 | 1259 | 96 | |
BC121894.1 | - | 2 - 3 | 1370 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |